missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ Toluene, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

ACS Reagent,≥99.5%

$299.19 - $957.97

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.141
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym antisal 1a
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60047300 View Documents Honeywell Riedel-de Haen™
6767564L
4L Aluminum bottle Each for $299.19
 
60047301 View Documents Honeywell Riedel-de Haen™
6767564X4L
4 x 4L Glass bottle Case of 4 Each for $957.97
 
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

108-88-3
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
antisal 1a
CHEBI:17578
CC1=CC=CC=C1

Specifications

108-88-3
Clear liquid
MFCD00008512
635760
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14g/mol
0.865g/mL at 25°C
997°C
3.46 (Air= 1.0)
106°C (1013hPa)
-140°C
Toluene
C7H8
C6H5CH3
UN1294
antisal 1a
CC1=CC=CC=C1
92.141
CHEBI:17578
≥99.5%
0.07
Aluminum bottle
59.9 hPa (at 25°C)
Undesignated
4L
Safety and Handling

Safety and Handling

P210-P261-P281-P301 + P310-P331

DOTInformation : class 3 - PG 2 - Toluene

RTECSNumber : 203-625-9