Please login to your online account to display your discounted pricing

Toluene, 99+%, for spectroscopy, ACROS Organics™

$70.11 - $123.96

Chemical Identifiers

Chemical Name or Material Toluene
Name Note 99+%
CAS 108-88-3
Purity Grade For spectroscopy
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7 H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC167900025

  SDS
thermo fisher scientific
167900025
Glass bottle 2.5L Each for $123.96

AC167900010

  SDS
thermo fisher scientific
167900010
Glass bottle 1L Each for $70.11
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥99.0%
Formula Weight 92.14
Quantity 2.5L
Color Clear
Beilstein 05, 280
Merck Index 15, 9688
EINECS Number 203-625-9
TSCA TSCA
Purity Grade For spectroscopy
Density 0.8660g/mL
RTECS Number XS5250000
Chemical Name or Material Toluene
Packaging Glass bottle
Absorbance (1 cm cell vs water)
Health Hazard 1 Signal Word:DANGER!
Additional Information Vapor Pressure: 29mbar at 20°C
Residue after Evaporation 0.001% max.
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C7 H8
Linear Formula C6H5CH3
Synonym Methylbenzene
Absorbance Conditions (1 cm cell vs water)
Water 0.03% max.
MDL Number MFCD00008512
CAS 108-88-3
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

For UV/VIS spectroscopy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Signal Word:DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

GHS H Statement:
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.