Please login to your online account to display your discounted pricing

Toluene, 99.85%, for residue analysis, for pesticide residue analysis, ACROS Organics™

$70.34 - $70.34

Chemical Identifiers

Chemical Name or Material Toluene
Name Note For Pesticide Residue Analysis
CAS 108-88-3
Purity Grade For residue analysis
Assay ≥99.80% (GC)
Assay Percent Range 99.85%
Molecular Formula C7H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC332070010

  SDS
thermo fisher scientific
332070010
Glass bottle 1L Each for $70.34
Description & Specifications

Specifications

Assay ≥99.80% (GC)
Physical Form Liquid
Formula Weight 92.14
Quantity 1L
Color Clear colorless to light yellow
Beilstein 05, 280
Merck Index 15, 9688
EINECS Number 203-625-9
TSCA TSCA
Water 0.02% Max. (K.F.)
Purity Grade For residue analysis
Density 0.8660g/mL
RTECS Number XS5250000
Additional Information Vapor Pressure: 29mbar at 20°C
Synonym Methylbenzene
Health Hazard 1 Signal Word:DANGER!
Health Hazard 2 GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Identification Pass Test
Packaging Glass bottle
Residue after Evaporation 0.0005 % max.
Assay Percent Range 99.85%
Acidity ≤0.003% (as HCl)
Hydrochloric Acid 0.003% max.
Chemical Name or Material Toluene
Name Note For Pesticide Residue Analysis
Molecular Formula C7H8
Linear Formula C6H5CH3
MDL Number MFCD00008512
CAS 108-88-3

For residue analysis

Safety and Handling

Signal Word:DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag