Please login to your online account to display your discounted pricing

Toluene, 99.85%, for HPLC, ACROS Organics™

$73.92 - $156.47

Chemical Identifiers

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade For HPLC
Assay ≥99.8%
Assay Percent Range 99.85%
Molecular Formula C7 H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Synonym Methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC268370010

  SDS
thermo fisher scientific
268370010
Glass bottle 1L Each for $73.92

AC268370025

  SDS
thermo fisher scientific
268370025
Glass bottle 2.5L Each for $156.47
Description & Specifications

Specifications

Assay ≥99.8%
Physical Form Liquid
Formula Weight 92.14
Quantity 1L
Color Clear
Density 0.8660g/mL
RTECS Number XS5250000
TSCA TSCA
EINECS Number 203-625-9
Purity Grade For HPLC
Beilstein 05, 280
Merck Index 15, 9688
Absorbance (1 cm cell vs HPLC−grade water)
Residue after Evaporation 0.0002% max.
Filtered to 0.2 micrometer Yes
Synonym Methylbenzene
Absorbance Conditions (1 cm cell vs HPLC-grade water)
Additional Information Vapor Pressure: 29mbar at 20°C
Chemical Name or Material Toluene
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.
MDL Number MFCD00008512
Water 0.03% max.
Acidity 0.0015 meq/g max.
Packaging Glass bottle
Health Hazard 1 Signal Word:DANGER!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Linear Formula C6H5CH3
Molecular Formula C7 H8
Assay Percent Range 99.85%
CAS 108-88-3

For HPLC

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Signal Word:DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

GHS H Statement:
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.