Please login to your online account to display your discounted pricing

Toluene, 99.85%, Extra Dry, AcroSeal™, ACROS Organics™


Quantity 100mL

Manufacturer: thermo fisher scientific  326981000

 View more versions of this product

Catalog No. AC326981000

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Toluene
Name Note Extra Dry
CAS 108-88-3
Health Hazard 1 Signal Word:DANGER!
Health Hazard 2 GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Additional Information Vapor Pressure: 29mbar at 20°C
Purity Grade Extra Dry
Physical Form Liquid
Assay ≥99.8% (GC)
Assay Percent Range 99.85%
Molecular Formula C7H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Beilstein 05, 280
Merck Index 15, 9688
Synonym Methylbenzene
Density 0.8660g/mL
Hydrochloric Acid 0.002% max.
Infrared Spectrum Authentic
Residue after Evaporation 0.0005 % max.
Water 0.003% Max. (Coulometric)
Filtered Through Filtered to 0.2 micromete PFTE
Color Clear colorless
Quantity 100mL

Specifications

Chemical Name or Material Toluene
Name Note Extra Dry
CAS 108-88-3
Purity Grade Extra Dry
Assay ≥99.8% (GC)
Assay Percent Range 99.85%
Molecular Formula C7H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Synonym Methylbenzene
Accessories

Signal Word:DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag

SDS & Safety Documents