missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 610460010

 View more versions of this product

Catalog No. AC610460010



Specifications

Specifications

Toluene
108-88-3
99.8%
C6H5CH3
05, 280
antisal 1a
CC1=CC=CC=C1
92.141
CHEBI:17578
≥99.8%
0.8660g/mL
50ppm max.
Water < 50ppm
Vapor Pressure: 29mbar at 20°C
C7H8
MFCD00008512
15, 9688
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
Extra Dry
Pass Test
1L

Chemical Identifiers

108-88-3
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
antisal 1a
CHEBI:17578
CC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

Signal Word: DANGER!

EINECSNumber : 203-625-9

RTECSNumber : XS5250000

TSCA : TSCA

SDS Documents