missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene, 99.7% min, HPLC Grade, Alfa Aesar™

Manufacturer:  Alfa Aesar 22903M1

 View more versions of this product

Catalog No. AA22903M1



Specifications

Specifications

Toluene
HPLC
C7H8
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CC1=CC=CC=C1
92.141
CHEBI:17578
Liquid
4 x 1L
108-88-3
99.7% min.
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
0.867

Chemical Identifiers

108-88-3
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Safety and Handling

Safety and Handling

EINECSNumber : 203-625-9

SDS Documents