Please login to your online account to display your discounted pricing

Toluene, 99.5%, for spectroscopy ACS, ACROS Organics™


Quantity 25L
Packaging Metal drum

Manufacturer: thermo fisher scientific  AC421170250

 View more versions of this product

Catalog No. AC421170250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damaging the unborn child. Highly flammable liquid and vapor.
Assay ≥99.5%
Physical Form Liquid
Formula Weight 92.14
Purity Grade ACS Grade
Quantity 25L
Color Clear
Beilstein 05, 280
Merck Index 15, 9688
TSCA TSCA
EINECS Number 203-625-9
RTECS Number XS5250000
Density 0.8660g/mL
Sulfur Compounds 0.003% max.
Absorbance 0.01 max. at 350nm
Substances Darkened by H2SO4 Passes Test
Molecular Formula C7H8
Name Note 99.5%
Assay Percent Range 99.5%
MDL Number MFCD00008512
Residue after Evaporation 0.001% max.
Water 0.03% max. (Coulometric)
Synonym Methylbenzene
Packaging Metal drum
Additional Information Vapor Pressure: 29mbar at 20°C
Chemical Name or Material Toluene
CAS 108-88-3
Linear Formula C6H5CH3

Specifications

Chemical Name or Material Toluene
Name Note 99.5%
CAS 108-88-3
Purity Grade ACS Grade
Assay ≥99.5%
Assay Percent Range 99.5%
Molecular Formula C7H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Synonym Methylbenzene
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS H Statement
May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damaging the unborn child. Highly flammable liquid and vapor.

SDS & Safety Documents