Please login to your online account to display your discounted pricing

Toluene, 99.5%, ACS reagent, ACROS Organics™


Quantity 25mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  424550250

 View more versions of this product

Catalog No. AC424550250

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damaging the unborn child. Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.
Assay ≥99.5%
Physical Form Liquid
Formula Weight 92.14
Purity Grade ACS Reagent Grade
Quantity 25mL
Color Clear
Merck Index 15, 9688
Beilstein 05, 280
Packaging Glass Bottle
Density 0.8660g/mL
Linear Formula C6H5CH3
CAS 108-88-3
MDL Number MFCD00008512
Assay Percent Range 99.5%
Chemical Name or Material Toluene
Additional Information Vapor Pressure: 29mbar at 20°C
Sulfur Compounds 0.003% max.
Water 0.03% max. (Coulometric)
Residue after Evaporation 0.001% max.
Substances Darkened by H2SO4 Passes Test
Synonym Methylbenzene
Molecular Formula C7H8

Specifications

Chemical Name or Material Toluene
CAS 108-88-3
Purity Grade ACS Reagent Grade
Assay ≥99.5%
Assay Percent Range 99.5%
Molecular Formula C7H8
Linear Formula C6H5CH3
Formula Weight 92.14
MDL Number MFCD00008512
Synonym Methylbenzene
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS H Statement
May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damaging the unborn child. Highly flammable liquid and vapor.

SDS & Safety Documents