missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Toluene-2,6-diisocyanate, 97%

Manufacturer:  Alfa Aesar™ L1274503

 View more versions of this product

Catalog No. AAL1274503



Description

Description

Applications
Toluene-2,6-diisocyanate is used as an organic chemical synthesis intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines and heat.
Specifications

Specifications

Toluene-2,6-diisocyanate
Pungent
13°C
91-08-7
MFCD00002010
2211546
RUELTTOHQODFPA-UHFFFAOYSA-N
1,3-diisocyanato-2-methylbenzene
7040
174.16
>110°C (230°F)
129°C to 133°C (18mmHg)
1g
C9H6N2O2
UN2078
2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene
CC1=C(C=CC=C1N=C=O)N=C=O
174.159
CHEBI:53557
97%

Chemical Identifiers

91-08-7
174.159
RUELTTOHQODFPA-UHFFFAOYSA-N
7040
1,3-diisocyanato-2-methylbenzene
C9H6N2O2
MFCD00002010
2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene
CHEBI:53557
CC1=C(C=CC=C1N=C=O)N=C=O
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H351-H315-H319-H317-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P273-P305+P351+P338-P304+P340-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUENE DIISOCYANATE

RTECSNumber : CZ6310000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents