Please login to your online account to display your discounted pricing

Tolterodine L-tartrate, Tocris™

Potent, non-selective muscarinic receptor antagonist

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Tolterodine L-tartrate
Assay Percent Range >99%
Molecular Formula C22H31NO·C4H6O6
Formula Weight 475.57
Synonym 2-[(1R)-3-[Bis(1-methylethyl)amino]-1-phenylpropyl]-4-methylphenol (2R, 3R)-2, 3-dihydroxybutanedioate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

376110

 
r & d systems
RDS376110
10mg Each for $164.61

376150

 
r & d systems
RDS376150
50mg Each for $682.67
Description & Specifications

Specifications

Formula Weight 475.57
Quantity 10mg
Recommended Storage Store at +4°C
Molecular Formula C22H31NO·C4H6O6
Assay Percent Range >99%
CAS 124937-52-6
Chemical Name or Material Tolterodine L-tartrate
Synonym 2-[(1R)-3-[Bis(1-methylethyl)amino]-1-phenylpropyl]-4-methylphenol (2R, 3R)-2, 3-dihydroxybutanedioate
Safety and Handling

Store at +4°C