Please login to your online account to display your discounted pricing

Thermo Scientific™ Todd Hewitt Broth with Gentamicin and Nalidixic Acid (Trans Vag Broth)

Reduce media prep time while selectively isolating group B streptococci from clinical specimens containing mixed flora with pre-filled, ready-to-use tubes.

$519.10 - $519.10

Specifications

Color Clear liquid, light amber
Description Todd Hewitt Broth with Gentamicin and Nalidixic Acid (Trans Vag Broth)
Format Tubes
Fill Volume 5mL
Manufacturing Origin United States
View More Specs
Products
Catalog Number Mfr. No. Description Manufacturing Origin Format Quantity Price Quantity    

R064842

 
thermo scientific™
RE064842
Todd Hewitt Broth with Gentamicin and Nalidixic Acid (Trans Vag Broth) United States Tubes 20/pk. Pack of 20 Each for $519.10
Description & Specifications

Specifications

Color Clear liquid, light amber
Description Todd Hewitt Broth with Gentamicin and Nalidixic Acid (Trans Vag Broth)
Format Tubes
Fill Volume 5mL
Manufacturing Origin United States
Storage Requirements 2-8°C

Selectively isolate group B streptococci from clinical specimens containing mixed flora using Thermo Scientific™ Todd Hewitt Broth with Gentamicin and Nalidixic Acid (Trans Vag Broth).

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Todd Hewitt Broth with the addition of Gentamicin and Nalidixic Acid to inhibit growth of some gram-negative bacteria. Requires the addition of 0.25mL of 5% sheep blood per tube before use to increase recovery of group B streptococci.

Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.


Not all products are available for sale in all territories. Please inquire.


Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.