Please login to your online account to display your discounted pricing

Tocris ICI 174,864


δ selective peptide antagonist

Manufacturer: r & d systems  RDS08201

 View more versions of this product

Catalog No. 08201

  • / Each
 

Description & Specifications

Specifications

Quantity 1mg
Molecular Weight 691.87
Purity purity determined by batch