missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tobramycin sulfate, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
$293.21 - $859.12
Chemical Identifiers
CAS | 79645-27-5 |
---|---|
Molecular Formula | C18H39N5O13S |
Molecular Weight (g/mol) | 565.59 |
MDL Number | MFCD00133864,MFCD00133864 |
InChI Key | ZEUUPKVZFKBXPW-UHFFFAOYNA-N |
PubChem CID | 54413 |
IUPAC Name | 4-amino-2-[(4,6-diamino-3-{[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-6-(hydroxymethyl)oxane-3,5-diol; sulfuric acid |
SMILES | OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(N)CC1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67340XF
|
Thermo Scientific Chemicals
J67340XF |
1 mL |
Each for $293.21
|
|
|||||
AAJ673408EQ
|
Thermo Scientific Chemicals
J673408EQ |
5 x 1 mL |
Pack for $859.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79645-27-5 | |
565.59 | |
ZEUUPKVZFKBXPW-UHFFFAOYNA-N | |
4-amino-2-[(4,6-diamino-3-{[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-6-(hydroxymethyl)oxane-3,5-diol; sulfuric acid |
C18H39N5O13S | |
MFCD00133864,MFCD00133864 | |
54413 | |
OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(N)CC1O |
Specifications
Tobramycin Sulfate | |
79645-27-5 | |
MFCD00133864,MFCD00133864 | |
ZEUUPKVZFKBXPW-UHFFFAOYNA-N | |
4-amino-2-[(4,6-diamino-3-{[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-6-(hydroxymethyl)oxane-3,5-diol; sulfuric acid | |
54413 | |
Tobramycin sulfate |
Liquid | |
C18H39N5O13S | |
Plastic Bottle | |
OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(N)CC1O | |
565.59 | |
1 mL |
Safety and Handling
H335
RTECSNumber : WK2110000
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only