Please login to your online account to display your discounted pricing

Alfa Aesar™ Tobramycin sulfate, 50mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Tobramycin sulfate
CAS 79645-27-5
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00133864
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ673408EQ

 
alfa aesar™
J673408EQ
5 x 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00133864
CAS 79645-27-5
Name Note 50mg/mL in distilled water, sterile-filtered
Physical Form Liquid
Storage Note 1 -30° to -10°C
RTECS Number WK2110000
Chemical Name or Material Tobramycin sulfate
Quantity 5 x 1mL
Safety and Handling

-30° to -10°C