missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TNT buffer soln.

Specifications

Chemical Name or Material TNT buffer soln.
Synonym TRIS NaCl Tween 20 buffer soln.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60622K2 View Documents Alfa Aesar™
J60622K2
1L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ60622K3 View Documents Alfa Aesar™
J60622K3
2L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

TNT buffer soln.
TRIS NaCl Tween 20 buffer soln.