missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNF alpha Rat anti-Mouse, eFluor 450, Clone: MP6-XT22, eBioscience™

Rat Monoclonal Antibody

$172.55 - $368.30

Specifications

Antigen TNF alpha
Clone MP6-XT22
Host Species Rat
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124216 Life Technologies
LS48732182
100μg Each for $368.30
5016403 Life Technologies
LS48732180
25μg Each for $172.55
Specifications

Description

The MP6-XT22 antibody reacts with mouse tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4(+) T cells. TNF alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane-bound form.
Description & Specifications

Specifications

TNF alpha
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MP6-XT22
Tumor Necrosis Factor alpha
Flow Cytometry (Intracellular Staining)
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL