missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNF alpha Rat anti-Mouse, Clone: MP6-XT3, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$130.68 - $368.28

Specifications

Antigen TNF alpha
Clone MP6-XT3
Host Species Rat
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122999 Life Technologies
LS16732285
500μg Each for $368.28
5014752 Life Technologies
LS16732281
50μg Each for $130.68
Specifications

Description

The MP6-XT3 monoclonal antibody reacts with mouse Tumor Necrosis Factor-alpha (TNF-α), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4+ T cells. TNF alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane-bound form.
Description & Specifications

Specifications

TNF alpha
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, no sodium azide
MP6-XT3
Tumor Necrosis Factor alpha
Cytokine Neutralization
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL