Please login to your online account to display your discounted pricing

TNF alpha Rat anti-Mouse, Alexa Fluor 488, Clone: MP6-XT22, eBioscience™

Rat Monoclonal Antibody

$158.05 - $361.05

Specifications

Antigen TNF alpha
Clone MP6-XT22
Host Species Rat
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016810

 
life technologies
LS53732180
25μg Each for $158.05

5016811

 
life technologies
LS53732182
100μg Each for $361.05
Description & Specifications

Specifications

Antigen TNF alpha
Clone MP6-XT22
Host Species Rat
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MP6-XT22 antibody reacts with mouse tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4(+) T cells. TNF alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane-bound form.