Please login to your online account to display your discounted pricing

TNF alpha Mouse anti-Human, PE-eFluor 610, Clone: MAb11, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $477.05

Specifications

Antigen TNF alpha
Clone MAb11
Host Species Mouse
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124878

 
life technologies
LS61734941
25 tests Each for $187.05

501124879

 
life technologies
LS61734942
100 tests Each for $477.05
Description & Specifications

Specifications

Antigen TNF alpha
Clone MAb11
Host Species Mouse
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The MAb11 antibody reacts with human tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4+ T cells. TN alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane bound form. The MAb11 antibody is a neutralizing antibody.