missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNF alpha Mouse anti-Human, FITC, Clone: MAb11, eBioscience™

Mouse Monoclonal Antibody

$118.80 - $302.28

Specifications

Antigen TNF alpha
Clone MAb11
Host Species Murine
Gene Alias Tumor Necrosis Factor alpha
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010037 Life Technologies
LS11734941
25 Tests Each for $118.80
5010038 Life Technologies
LS11734981
50μg Each for $174.24
5010039 Life Technologies
LS11734982
100μg Each for $302.28
Specifications

Description

The MAb11 antibody reacts with human tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4+ T cells. TN alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane bound form. The MAb11 antibody is a neutralizing antibody.
Description & Specifications

Specifications

TNF alpha
Murine
Flow Cytometry (Intracellular Staining)
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL; 5μL (0.5μg)/test
MAb11
Tumor Necrosis Factor alpha
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer