Please login to your online account to display your discounted pricing

TNF alpha Mouse anti-Human, eFluor 450, Clone: MAb11, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $442.25

Specifications

Antigen TNF alpha
Clone MAb11
Host Species Murine
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016405

 
life technologies
LS48734941
25 Tests Each for $216.05

5016406

 
life technologies
LS48734942
100 Tests Each for $442.25
Description & Specifications

Specifications

Antigen TNF alpha
Clone MAb11
Host Species Murine
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The MAb11 antibody reacts with human tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4+ T cells. TN alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane bound form. The MAb11 antibody is a neutralizing antibody.