missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNF alpha Mouse anti-Human, Clone: MAb1, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$130.68 - $368.28

Specifications

Antigen TNF alpha
Clone MAb1
Host Species Murine
Gene Alias Tumor Necrosis Factor alpha, TNFa TNF-a
Applications Cytokine Neutralization,ELISA Capture,ELISPOT Capture Antibody
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014758 Life Technologies
LS16734881
50μg Each for $130.68
5014759 Life Technologies
LS16734885
500μg Each for $368.28
Specifications

Description

The MAb1 antibody reacts with human tumor necrosis factor-alpha (TNF-α), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4+ T cells. TNF-α has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF-α forms dimers and trimers and also exists as a 26kDa membrane bound form.
Description & Specifications

Specifications

TNF alpha
Murine
Cytokine Neutralization,ELISA Capture,ELISPOT Capture Antibody
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
MAb1
Tumor Necrosis Factor alpha, TNFa TNF-a
RUO
Purified
Primary
aqueous buffer, no sodium azide