missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNF alpha Mouse anti-Human, Biotin, Clone: MAb11, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen TNF alpha
Clone MAb11
Host Species Murine
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012211 Life Technologies
LS13734981
50μg Each for $143.55
5012212 Life Technologies
LS13734985
500μg Each for $361.05
Specifications

Description

The MAb11 antibody reacts with human tumor necrosis factor-alpha (TNF-α ), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4+ T cells. TNF-α has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF-α forms dimers and trimers and also exists as a 26kDa membrane bound form. The MAb11 antibody is a neutralizing antibody.
Description & Specifications

Specifications

TNF alpha
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MAb11
Tumor Necrosis Factor alpha
ELISA Detection,ELISPOT Detection Antibody
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL