Please login to your online account to display your discounted pricing

TNF alpha Mouse anti-Human, APC, Clone: MAb11, eBioscience™

Mouse Monoclonal Antibody

$162.40 - $390.05

Specifications

Antigen TNF alpha
Clone MAb11
Host Species Mouse
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015328

 
life technologies
LS17734941
25 tests Each for $162.40

5015329

 
life technologies
LS17734982
100μg Each for $390.05
Description & Specifications

Specifications

Antigen TNF alpha
Clone MAb11
Host Species Mouse
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL; 5μL (0.125μg)/test

The MAb11 antibody reacts with human tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4+ T cells. TN alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane bound form. The MAb11 antibody is a neutralizing antibody.