Please login to your online account to display your discounted pricing

TNF alpha Armenian Hamster anti-Mouse, Rat, Clone: TN3-19.12, Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody, Functional Grade

$143.55 - $2,898.55

Specifications

Antigen TNF alpha
Clone TN3-19.12
Host Species Armenian Hamster
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Mouse, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123003

 
life technologies
LS16742338
5mg Each for $2,898.55

501123004

 
life technologies
LS16742385
500μg Each for $404.55

5014760

 
life technologies
LS16742381
50μg Each for $143.55
Description & Specifications

Specifications

Antigen TNF alpha
Clone TN3-19.12
Host Species Armenian Hamster
Gene Alias Tumor Necrosis Factor alpha
Species Reactivity Mouse, Rat
Applications Cytokine Neutralization
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The TN3-19.12 monoclonal antibody reacts with mouse, rat, and rabbit tumor necrosis factor alpha (TNF alpha), but not with human TNF alpha. The TN3-19.12 antibody is a neutralizing antibody. Mouse TNF alpha is a 17kDa factor produced by macrophages, monocytes, neutrophils, CD4+ T cells and NK cells. A 26kDa form of TNF alpha is expressed as a membrane-bound molecule. TNF alpha is cytolytic and plays an important role in immune regulation. Dimers and trimers of TNF alpha have been observed.