missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNBT Diformazan, TCI America™

$111.53 - $333.06

Chemical Identifiers

CAS 19333-63-2
Molecular Formula C40H30N12O10
Molecular Weight (g/mol) 838.754
MDL Number MFCD00059831
InChI Key SDKSRZQMARNUEW-DBPDIXJWSA-N
Synonym 1,1′C-(3,3′C-Dimethoxy-4,4′C-diphenylene)bis-3,5-di(p-nitrophenyl)diformazan, Tetranitro Blue Tetrazolium Diformazan
PubChem CID 44630091
IUPAC Name N-[2-methoxy-4-[3-methoxy-4-[[(Z)-N-(4-nitroanilino)-C-(4-nitrophenyl)carbonimidoyl]diazenyl]phenyl]phenyl]imino-4-nitro-N'-(4-nitroanilino)benzenecarboximidamide
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N=NC(=NNC3=CC=C(C=C3)[N+](=O)[O-])C4=CC=C(C=C4)[N+](=O)[O-])OC)N=NC(=NNC5=CC=C(C=C5)[N+](=O)[O-])C6=CC=C(C=C6)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B0282100MG View Documents TCI America
B0282100MG
100mg Each for $111.53
 
B02821G View Documents TCI America
B02821G
1g Each for $333.06
 
Specifications

Chemical Identifiers

19333-63-2
838.754
SDKSRZQMARNUEW-DBPDIXJWSA-N
44630091
COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N=NC(=NNC3=CC=C(C=C3)[N+](=O)[O-])C4=CC=C(C=C4)[N+](=O)[O-])OC)N=NC(=NNC5=CC=C(C=C5)[N+](=O)[O-])C6=CC=C(C=C6)[N+](=O)[O-]
C40H30N12O10
MFCD00059831
1,1′C-(3,3′C-Dimethoxy-4,4′C-diphenylene)bis-3,5-di(p-nitrophenyl)diformazan, Tetranitro Blue Tetrazolium Diformazan
N-[2-methoxy-4-[3-methoxy-4-[[(Z)-N-(4-nitroanilino)-C-(4-nitrophenyl)carbonimidoyl]diazenyl]phenyl]phenyl]imino-4-nitro-N'-(4-nitroanilino)benzenecarboximidamide

Specifications

Undesignated
19333-63-2
MFCD00059831
1,1′C-(3,3′C-Dimethoxy-4,4′C-diphenylene)bis-3,5-di(p-nitrophenyl)diformazan, Tetranitro Blue Tetrazolium Diformazan
COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N=NC(=NNC3=CC=C(C=C3)[N+](=O)[O-])C4=CC=C(C=C4)[N+](=O)[O-])OC)N=NC(=NNC5=CC=C(C=C5)[N+](=O)[O-])C6=CC=C(C=C6)[N+](=O)[O-]
838.754
838.75
TNBT Diformazan
100mg
C40H30N12O10
1325
SDKSRZQMARNUEW-DBPDIXJWSA-N
N-[2-methoxy-4-[3-methoxy-4-[[(Z)-N-(4-nitroanilino)-C-(4-nitrophenyl)carbonimidoyl]diazenyl]phenyl]phenyl]imino-4-nitro-N'-(4-nitroanilino)benzenecarboximidamide
44630091
Crystal-Powder at 20°C
Safety and Handling

Safety and Handling

TSCA : No