missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TN buffer soln.

Specifications

Chemical Name or Material TN buffer soln.
Synonym TRIS NaCl buffer soln
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63985AP View Documents Alfa Aesar™
J63985AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ63985K2 View Documents Alfa Aesar™
J63985K2
1L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

TN buffer soln.
TRIS NaCl buffer soln