Please login to your online account to display your discounted pricing

TMS, Tocris

Cytochrome P450 1B1 inhibitor

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material TMS
Assay Percent Range >98%
Molecular Formula C18H20O4
Formula Weight 300.35
Synonym 1-[2,(3,5-Dimethoxyphenyl)ethenyl]-2,4-dimethoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

150910

 
r & d systems
RDS150910
10mg Each for $225.77

150950

 
r & d systems
RDS150950
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 300.35
Quantity 10mg
Chemical Name or Material TMS
Molecular Formula C18H20O4
Assay Percent Range >98%
CAS 24144-92-1
Synonym 1-[2,(3,5-Dimethoxyphenyl)ethenyl]-2,4-dimethoxybenzene
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature