Please login to your online account to display your discounted pricing

TMPH hydrochloride, Tocris

Neuronal nicotinic receptor antagonist

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material TMPH hydrochloride
Assay Percent Range >98%
Molecular Formula C16H31NO2·HCl
Formula Weight 305.88
Synonym 2,2,6,6-Tetramethylpiperidin-4-yl heptanoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

243850

 
r & d systems
RDS243850
50mg Each for $682.67

243810

 
r & d systems
RDS243810
10mg Each for $164.61
Description & Specifications

Specifications

Formula Weight 305.88
Quantity 50mg
Synonym 2,2,6,6-Tetramethylpiperidin-4-yl heptanoate
Recommended Storage Desiccate at Room Temperature
Molecular Formula C16H31NO2·HCl
Assay Percent Range >98%
Chemical Name or Material TMPH hydrochloride
CAS 849461-91-2
Safety and Handling

Desiccate at Room Temperature