Please login to your online account to display your discounted pricing

TMN 355, Tocris™

Cyclophilin A inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material TMN 355
Assay Percent Range >98%
Molecular Formula C21H14ClFN2O2
Formula Weight 380.8
Synonym 2-Chloro-N-[(9H-fluoren-9-ylamino)carbonyl]-6-fluorobenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

415210

 
r & d systems
RDS415210
10mg Each for $164.61

415250

 
r & d systems
RDS415250
50mg Each for $682.67
Description & Specifications

Specifications

Formula Weight 380.8
Quantity 10mg
Assay Percent Range >98%
Synonym 2-Chloro-N-[(9H-fluoren-9-ylamino)carbonyl]-6-fluorobenzamide
Recommended Storage Store at Room Temperature
CAS 1186372-20-2
Molecular Formula C21H14ClFN2O2
Chemical Name or Material TMN 355
Safety and Handling

Store at Room Temperature