missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TM2 TEAD inhibitor

Catalog No. 791710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
791710 10 mg
791750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 791710 Supplier Tocris Bioscience™ Supplier No. 7917/10
Only null left

Potent and reversible inhibitor of TEAD mediated Hippo/Yap signalling

TM2 TEAD inhibitor is a potent and reversible TEA domain transcription factor inhibitor (IC50 = 38 nM and 156 nM for TEAD4 auto-palmitoylation and TEAD2 palmitoylation, respectively). It inhibits TEAD-YAP association and transcriptional activity. TM2 TEAD inhibitor exhibits strong antiproliferation effects as a single agent or in combination with a MEK inhibitor in YAP-dependent cancer cells. Also inhibits YAP-dependent liver organoid growth ex vivo.

Specifications

Chemical Name or Material 4-[3-(2-Cyclohexylethoxy)benzoyl]-N-phenyl-1-piperazinecarboxamide
CAS 1008768-41-9
Quantity 10 mg
Target YAP1
Molecular Formula C26H33N3O3
Purity 0.98