missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TLR4/MD2 Rat anti-Mouse, PE, Clone: MTS510, eBioscience™

Rat Monoclonal Antibody

$249.48 - $381.48

Specifications

Antigen TLR4/MD2
Clone MTS510
Host Species Rat
Gene Alias TLR4/ MD-2, TLR-4/ MD2, toll-like receptor MD2 complex
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122492 Life Technologies
LS12992482
100μg Each for $381.48
5011295 Life Technologies
LS12992481
50μg Each for $249.48
Specifications

Description

The MTS510 monoclonal antibody reacts with the mouse Toll-like receptor 4 (TLR4)/MD-2 complex. At least ten members of the Toll family have been identified. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR4 physically associates with MD-2, and together with CD14, this complex is responsible for LPS recognition and signaling. In the mouse, TLR4 is expressed by thioglycolate-elicited peritoneal macrophages. Incubation of peritoneal macrophages with LPS results in down regulation of surface TLR4/MD-2. The TLR4 gene is defective in C3H/HeJ and C57BL/10ScCr mice, both of which have been well characterized as hyporesponders to LPS.

The MTS510 monoclonal antibody co-immunoprecipitates MD-2 (∽30 kDa) and TLR4 (∽100 kDa), and preferentially reacts with TLR4 that is associated with MD-2. In comparison, binding of the UT41 monoclonal antibody occurs with and without formation of the TLR4/MD-2 complex . Please contact eBioscience Technical Support for further information.
Description & Specifications

Specifications

TLR4/MD2
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MTS510
TLR4/ MD-2, TLR-4/ MD2, toll-like receptor MD2 complex
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL