missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Human TLK1, GST Tag Recombinant Protein DFS Item

TLK1 or tousled-like kinase 1 is a nuclear serine/threonine kinase that is potentially involved in the regulation of chromatin assembly. Recombinant full-length human TLK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

Supplier:  Invitrogen™ PV6422

 View more versions of this product

Catalog No. PV6422



Description

Description

TLK1 is first described in Arabidopsis. TLK1 is a novel target of the DNA damage checkpoint that is rapidly inactivated upon exposure to ionizing radiation (IR) and the inactivation is directly mediated by the S-phase DNA damage checkpoint. The suppression of TLK1 activity after double-strand breaks in the DNA, replication blockade, or low doses of ultraviolet irradiation requires ATM, NBS and CHK1.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
113.0kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
100μg
KIAA0137, PKU-beta
Recombinant
SDS
Documents

Documents

Promotions

Promotions