missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tizanidine Hydrochloride, TCI America™

Manufacturer:  TCI America T252725G

 View more versions of this product

Catalog No. T252725G



Specifications

Specifications

Tizanidine Hydrochloride
280°C
64461-82-1
MFCD00798231
ZWUKMNZJRDGCTQ-UHFFFAOYSA-N
5-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine;hydrochloride
114869
Crystal-Powder at 20°C
White-Yellow
25g
C9H9Cl2N5S
5-Chloro-4-[(4,5-dihydro-1H-imidazol-2-yl)amino]-2,1,3-benzothiadiazole Hydrochloride
C1CN=C(N1)NC2=C(C=CC3=NSN=C32)Cl.Cl
290.166
290.17
≥98.0% (HPLC,T)

Chemical Identifiers

64461-82-1
290.166
ZWUKMNZJRDGCTQ-UHFFFAOYSA-N
114869
C1CN=C(N1)NC2=C(C=CC3=NSN=C32)Cl.Cl
C9H9Cl2N5S
MFCD00798231
5-Chloro-4-[(4,5-dihydro-1H-imidazol-2-yl)amino]-2,1,3-benzothiadiazole Hydrochloride
5-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine;hydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : DK9910000

TSCA : No

Documents