Please login to your online account to display your discounted pricing

Titration (TAN) Solvent, Certified, LabChem™

$210.50 - $210.50

Chemical Identifiers

Chemical Name or Material Titration (TAN) Solvent
CAS 108-88-3
CAS Max % 50
CAS 67-63-0
CAS Max % 49.5
CAS 7732-18-5
CAS Max % 0.5
Purity Grade Certified Grade
Linear Formula C7H8
Formula Weight 92.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC26140-4

 
lab chem
LC261404
Amber Glass 4L Each for $210.50
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 92.14
Chemical Name or Material Titration (TAN) Solvent
CAS Max % 0.5
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
Color Clear, colorless
Quantity 4L
Physical Form Liquid
Linear Formula C7H8
CAS 108-88-3
UN Number UN1993
Health Hazard 1 Danger
CAS Max % 50
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
May cause damage to organs (central nervous system, bone marrow, liver, kidneys) through prolonged or repeated exposure (oral, inhalation).
Harmful to aquatic life.
Identification Passes Test
CAS 67-63-0
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Packaging Amber Glass
CAS Max % 49.5
Solubility Soluble in water
CAS 7732-18-5
Suitability Passes Test
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
May cause damage to organs (central nervous system, bone marrow, liver, kidneys) through prolonged or repeated exposure (oral, inhalation).
Harmful to aquatic life.