missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium wire, 0.25mm dia., 99.7% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar™ 10396H2

 View more versions of this product

Catalog No. AA10396H2



Specifications

Specifications

Titanium
7440-32-6
99.7% (Metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
50m
Silver Gray wire
Ti
alloy, c.p., contimet 30, cp, ii hydride, jistp28, oremet, titan vt 1-1, titanium-125, vt1
[Ti]
Odorless
23963
Wire, ≈0.22g/m,Temper: as drawn

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy, c.p., contimet 30, cp, ii hydride, jistp28, oremet, titan vt 1-1, titanium-125, vt1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

EINECSNumber : 231-142-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents