missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium wire, 99.5% (metals basis), Alfa Aesar™

Temper: as drawn

Manufacturer:  Alfa Aesar 10391G5

 View more versions of this product

Catalog No. AA10391G5



Specifications

Specifications

Titanium
7440-32-6
4.506g/cm3
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
Odorless
23963
Wire, 0.127mm (0.005 in.) dia.
5m
Silver Gray wire
99.99% (Metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
Silver to Gray

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

EINECSNumber : 231-142-3

Documents