Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium wire, 2mm dia., 99.7% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Titanium
CAS 7440-32-6
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10397KJ

  SDS
alfa aesar™
10397KJ
5 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10397KM

  SDS
alfa aesar™
10397KM
25 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Wire, 2.0mm (0.08 in.) dia. X 91cm (36 in.) long
Quantity 5 pieces
EINECS Number 231-142-3
Odor Odorless
Chemical Name or Material Titanium
Appearance Silver Gray wire
Density 4.506g/cm3
Assay Percent Range 99.7% (Metals basis)
MDL Number MFCD00011264
CAS 7440-32-6