missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Titanium Wire, 2.0mm (0.08 in.) dia., 99.99% (Metals basis), Alfa Aesar™
Form: Approx. 14.2g/m; Temper: as drawn
Manufacturer: Alfa Aesar 14004BY
Specifications
Titanium | |
7440-32-6 | |
4.506g/cm3 | |
Ti | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
[Ti] | |
Odorless | |
23963 | |
Wire, 2mm (0.08 in.) dia. |
100cm | |
Silver Gray wire | |
99.99% (Metals basis) | |
MFCD00011264 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
titanium | |
47.867 | |
CHEBI:33341 | |
Silver to Gray |
Chemical Identifiers
7440-32-6 | |
47.867 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
23963 | |
titanium |
Ti | |
MFCD00011264 | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
CHEBI:33341 | |
[Ti] |