missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium Wire, 2.0mm (0.08 in.) dia., 99.99% (Metals basis), Alfa Aesar™

Form: Approx. 14.2g/m; Temper: as drawn

Manufacturer:  Alfa Aesar 14004BU

 View more versions of this product

Catalog No. AA14004BU



Specifications

Specifications

Titanium
7440-32-6
99.99% (Metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
Silver to Gray
25cm
Silver Gray wire
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
Odorless
23963
Wire, 2mm (0.08 in.) dia.

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Documents