missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium wire, 1.0mm (0.04 in.) dia., hard, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 45485BU

 View more versions of this product

Catalog No. AA45485BU



Specifications

Specifications

Titanium wire
25cm
7440-32-6
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
Odorless
titanium
23963
Wire
≈3.54g/m
1.0mm (0.04 in.) dia., Hard
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
[Ti]
47.867
CHEBI:33341
99.99%

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents