missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium Wire, 1.0mm (0.04 in.) dia., annealed, 99.99% (Metals basis), Alfa Aesar™

Form: Approx. 3.54g/m

Manufacturer:  Thermo Fisher Scientific 10388BU

 View more versions of this product

Catalog No. AA10388BU



Specifications

Specifications

Titanium
Annealed
Silver Gray wire
Ti
alloy
[Ti]
Odorless
23963
Wire, 1.0mm (0.04 in.) dia.
25cm
7440-32-6
99.99% (Metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
Silver to Gray

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Documents