missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium Thinfoil, 0.0114mm (0.00045 in.) thick, 99.6% (metals basis)

Manufacturer:  Alfa Aesar™ 44242GH

 View more versions of this product

Catalog No. AA44242GH



Specifications

Specifications

Titanium Thinfoil
100 x 100mm
7440-32-6
Ti
alloy
[Ti]
Odorless
23963
Thin Foil, ≈0.514g/100x100mm,Light Tight
Commercial Pure 4
≈0.514g/100x100mm
0.0114mm (0.00045 in.) thick, Light Tight
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
99.6%

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

EINECSNumber : 231-142-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents