missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium tablets, 2.5cm (1.0in) x 2.5cm (1.0in), 99+% (metals basis)

Manufacturer:  Alfa Aesar™ 45089P5

 View more versions of this product

Catalog No. AA45089P5



Specifications

Specifications

Titanium tablets
5 lb.
7440-32-6
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
Odorless
titanium
23963
≥99%
≈28.3g/tablet
2.5cm (1.0 in.) x 2.5cm (1.0 in.)
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
[Ti]
47.867
CHEBI:33341

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents