Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.7% (metals basis)

$446.00 - $446.00

Chemical Identifiers

Chemical Name or Material Titanium sputtering target
CAS 7440-32-6
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40905KS

  SDS
alfa aesar™
40905KS
1 each Each for $446.00
Description & Specifications

Specifications

Quantity 1 each
MDL Number MFCD00011264
Assay 99.7%
Odor Odorless
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Chemical Name or Material Titanium sputtering target
Assay Percent Notes (metals basis)
TSCA TSCA
CAS 7440-32-6