Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.995% (metals basis)

$620.32 - $620.32

Chemical Identifiers

Chemical Name or Material Titanium sputtering target
CAS 7440-32-6
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43044KS

 
alfa aesar™
43044KS
1 each Each for $620.32
Description & Specifications

Specifications

Chemical Name or Material Titanium sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.995%
Assay Percent Notes (metals basis)
MDL Number MFCD00011264
CAS 7440-32-6
Odor Odorless
Quantity 1 each