Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.995% (metals basis)

$593.66 - $593.66

Chemical Identifiers

Chemical Name or Material Titanium sputtering target
CAS 7440-32-6
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43043KS

  SDS
alfa aesar™
43043KS
1 each Each for $593.66
Description & Specifications

Specifications

MDL Number MFCD00011264
Assay 99.995%
CAS 7440-32-6
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Quantity 1 each
Chemical Name or Material Titanium sputtering target
Odor Odorless
Assay Percent Notes (metals basis)