Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.7% (metals basis)

$247.82 - $247.82

Chemical Identifiers

Chemical Name or Material Titanium sputtering target
CAS 7440-32-6
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40903KS

 
alfa aesar™
40903KS
1 each Each for $247.82
Description & Specifications

Specifications

Chemical Name or Material Titanium sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.7%
Assay Percent Notes (metals basis)
MDL Number MFCD00011264
CAS 7440-32-6
Odor Odorless
Quantity 1 each