Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.7% (metals basis)

Magnetron sputtering source

$423.41 - $423.41

Chemical Identifiers

Chemical Name or Material Titanium sputtering target
CAS 7440-32-6
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40902KS

  SDS
alfa aesar™
40902KS
1 each Each for $423.41
Description & Specifications

Specifications

Assay Percent Notes (metals basis)
Chemical Name or Material Titanium sputtering target
Assay 99.7%
Odor Odorless
Quantity 1 each
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
CAS 7440-32-6
TSCA TSCA
MDL Number MFCD00011264